Cyclopropyl(2,4-dimethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287061-1G 287061-500MG 287061-5G
77468-168EA 576.8 USD
77468-168 77480-860 77491-216
Cyclopropyl(2,4-dimethoxyphenyl)methanone ≥95%
Cyclopropyl(2,4-dimethoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14529970
CAS Number: 1216158-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR