5-(1,1-Dimethylethyl)-2-methoxy-α,α-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

287007-1G 287007-500MG 287007-5G
77468-160EA 676.71 USD
77468-160 77480-854 77491-208
5-(1,1-Dimethylethyl)-2-methoxy-α,α-dimethylbenzenemethanol ≥95%
5-(1,1-Dimethylethyl)-2-methoxy-α,α-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19351849
CAS Number: 1267278-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR