1-[5-(1,1-Dimethylethyl)-2-methoxyphenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

286994-1G 286994-500MG 286994-5G
77468-156EA 676.71 USD
77468-156 77480-850 77491-204
1-[5-(1,1-Dimethylethyl)-2-methoxyphenyl]-1-propanone ≥95%
1-[5-(1,1-Dimethylethyl)-2-methoxyphenyl]-1-propanone

Precaución: For research use only

Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14530326
CAS Number: 35155-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR