1-(3,5-Difluorophenyl)cyclopentan-1-ol ≥97%

Supplier: AOBChem USA

Warning

28612-1G 28612-500MG 28612-5G 28612-25G
77468-152EA 282.22 USD
77468-152 77480-846 77491-200 77501-230
1-(3,5-Difluorophenyl)cyclopentan-1-ol ≥97%
1-(3,5-Difluorophenyl)cyclopentan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O
MW: 198.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222251
CAS Number: 1344015-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR