4'-Formyl-2'-methyl[1,1'-biphenyl]-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

284861-1G 284861-500MG 284861-5G 284861-25G
77468-148EA 394.49 USD
77468-148 77480-842 77491-194 77501-224
4'-Formyl-2'-methyl[1,1'-biphenyl]-4-carboxylic acid ≥95%
4'-Formyl-2'-methyl[1,1'-biphenyl]-4-carboxylic acid

Precaución: For research use only

Formula: C₁₅H₁₂O₃
MW: 240.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547501
CAS Number: 1179362-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR