(6-Bromo-3-ethoxy-2-fluorophenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

284242-1G 284242-500MG 284242-5G 284242-25G
77468-140EA 415.09 USD
77468-140 77480-834 77491-186 77501-216
(6-Bromo-3-ethoxy-2-fluorophenyl)(cyclopropyl)methanol ≥95%
(6-Bromo-3-ethoxy-2-fluorophenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₄BrFO₂
MW: 289.1407 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34884963
CAS Number: 2969413-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR