1-(6-Bromo-2-fluoro-3-methoxyphenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

284240-1G 284240-500MG 284240-5G 284240-25G
77468-136EA 415.09 USD
77468-136 77480-830 77491-182 77501-212
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-3-methylbutan-1-ol ≥97%
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂
MW: 291.1566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34884961
CAS Number: 2969457-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR