1-(6-Bromo-3-chloro-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

284021-1G 284021-500MG 284021-5G 284021-25G
77468-132EA 204.97 USD
77468-132 77480-826 77491-178 77501-208
1-(6-Bromo-3-chloro-2-fluorophenyl)ethanol ≥95%
1-(6-Bromo-3-chloro-2-fluorophenyl)ethanol

Precaución: For research use only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD28141975
CAS Number: 1823493-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR