2-Chloro-6-isopropylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

28310-1G 28310-500MG 28310-5G 28310-25G
77468-128EA 339.9 USD
77468-128 77480-822 77491-174 77501-200
2-Chloro-6-isopropylnicotinaldehyde ≥97%
2-Chloro-6-isopropylnicotinaldehyde

Precaución: For research use only

Formula: C₉H₁₀ClNO
MW: 183.635 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22199711
CAS Number: 1289160-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR