Phenylmethyl 3-hydroxy-4-pentenoate ≥95%

Supplier: AOBChem USA

Warning

282386-1G 282386-500MG 282386-5G 282386-25G
77468-120EA 415.09 USD
77468-120 77480-814 77491-166 77501-194
Phenylmethyl 3-hydroxy-4-pentenoate ≥95%
Phenylmethyl 3-hydroxy-4-pentenoate

Precaución: For research use only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Storage Temperature: Ambient
CAS Number: 595569-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR