1-(2,3-Difluoro-4-methylphenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

280850-1G 280850-500MG 280850-5G 280850-25G
77468-108EA 284.28 USD
77468-108 77480-802 77491-154 77501-184
1-(2,3-Difluoro-4-methylphenyl)-3-methylbutan-1-ol ≥97%
1-(2,3-Difluoro-4-methylphenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O
MW: 214.2516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34883270
CAS Number: 1692557-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR