1-(5-Bromo-2-fluoro-3-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

280844-1G 280844-500MG 280844-5G 280844-25G
77468-104EA 415.09 USD
77468-104 77480-798 77491-150 77501-180
1-(5-Bromo-2-fluoro-3-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(5-Bromo-2-fluoro-3-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₀H₅BrF₆O
Storage Temperature: Ambient
MDL Number: MFCD34883265
CAS Number: 2969396-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR