3-(5-Bromo-2-fluoro-3-methylphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

280840-1G 280840-500MG 280840-5G 280840-25G
77468-100EA 415.09 USD
77468-100 77480-794 77491-146 77501-176
3-(5-Bromo-2-fluoro-3-methylphenyl)oxetan-3-ol ≥95%
3-(5-Bromo-2-fluoro-3-methylphenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34883261
CAS Number: 2969444-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR