1-(3-Chloro-2,6-difluorophenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

280798-1G 280798-500MG 280798-5G 280798-25G
77468-096EA 415.09 USD
77468-096 77480-790 77491-142 77501-172
1-(3-Chloro-2,6-difluorophenyl)-3-methylbutan-1-ol ≥95%
1-(3-Chloro-2,6-difluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34883244
CAS Number: 2969467-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR