Isopropyl 3-hydroxy-5-methylhexanoate ≥97%

Supplier: AOBChem USA

Warning

280794-1G 280794-500MG 280794-5G 280794-25G
77468-092EA 415.09 USD
77468-092 77480-786 77491-138 77501-168
Isopropyl 3-hydroxy-5-methylhexanoate ≥97%
Isopropyl 3-hydroxy-5-methylhexanoate

Precaución: For research use only

Formula: C₁₀H₂₀O₃
MW: 188.264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34883241
CAS Number: 2969394-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR