Isopropyl(2-methoxy-4-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

280400-1G 280400-500MG 280400-5G 280400-25G
77468-088EA 415.09 USD
77468-088 77480-782 77491-134 77501-164
Isopropyl(2-methoxy-4-methylphenyl)sulfane ≥95%
Isopropyl(2-methoxy-4-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34882920
CAS Number: 2969443-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR