2-Methoxy-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

27992-1G 27992-500MG 27992-5G 27992-25G
77468-084EA 221.45 USD
77468-084 77480-778 77491-130 77501-158
2-Methoxy-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%
2-Methoxy-4-(trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₄H₁₈BF₃O₄
MW: 318.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18760109
CAS Number: 2121514-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR