(2,6-Dichloro-4-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

27965-1G 27965-500MG 27965-5G 27965-25G
77468-080EA 195.7 USD
77468-080 77480-774 77491-126 77501-154
(2,6-Dichloro-4-(trifluoromethyl)phenyl)methanol ≥97%
(2,6-Dichloro-4-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₈H₅Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD27946698
CAS Number: 189338-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR