4-(4-(ethylthio)phenyl)tetrahydro-2H-pyran-4-ol ≥96%

Supplier: AOBChem USA

Warning

279639-1G 279639-500MG 279639-5G 279639-25G
77468-076EA 415.09 USD
77468-076 77480-770 77491-122 77501-150
4-(4-(ethylthio)phenyl)tetrahydro-2H-pyran-4-ol ≥96%
4-(4-(ethylthio)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₃H₁₈O₂S
MW: 238.3458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31923601
CAS Number: 2565807-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR