4'-(Ethylthio)-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

279635-1G 279635-500MG 279635-5G 279635-25G
77468-072EA 85.49 USD
77468-072 77480-766 77491-118 77501-148
4'-(Ethylthio)-2,2,2-trifluoroacetophenone ≥97%
4'-(Ethylthio)-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₁₀H₉F₃OS
Storage Temperature: Ambient
MDL Number: MFCD06201804
CAS Number: 845823-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR