1-(4-(ethylthio)phenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

279633-1G 279633-500MG 279633-5G 279633-25G
77468-068EA 369.77 USD
77468-068 77480-762 77491-114 77501-146
1-(4-(ethylthio)phenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(4-(ethylthio)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34882598
CAS Number: 1695606-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR