1-(4-(ethylthio)phenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

279625-1G 279625-500MG 279625-5G 279625-25G
77468-060EA 415.09 USD
77468-060 77480-754 77491-106 77501-138
1-(4-(ethylthio)phenyl)cyclobutanol ≥97%
1-(4-(ethylthio)phenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₆OS
MW: 208.3198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34882592
CAS Number: 2969245-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR