(3-Bromo-4-chloro-2-fluorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

279180-1G 279180-500MG 279180-5G 279180-25G
77468-056EA 415.09 USD
77468-056 77480-750 77491-102 77501-134
(3-Bromo-4-chloro-2-fluorophenyl)(methyl)sulfane ≥95%
(3-Bromo-4-chloro-2-fluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₇H₅BrClFS
Storage Temperature: Ambient
MDL Number: MFCD34882187
CAS Number: 2706908-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR