α-Methyl-4-pentylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

278908-1G 278908-500MG 278908-5G 278908-25G
77468-052EA 342.99 USD
77468-052 77480-746 77491-098 77501-130
α-Methyl-4-pentylbenzenemethanol ≥95%
α-Methyl-4-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00627976
CAS Number: 65189-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR