3,5-Dibromo-4-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

27888-1G 27888-500MG 27888-5G 27888-25G
77468-044EA 251.32 USD
77468-044 77480-740 77491-092 77501-126
3,5-Dibromo-4-isopropoxypyridine ≥95%
3,5-Dibromo-4-isopropoxypyridine

Precaución: For research use only

Formula: C₈H₉Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD26743624
CAS Number: 1357094-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR