2-(6-Bromo-3-ethoxy-2-fluorophenyl)pentan-2-ol ≥95%

Supplier: AOBChem USA

Warning

278798-1G 278798-500MG 278798-5G 278798-25G
77468-040EA 415.09 USD
77468-040 77480-734 77491-086 77501-120
2-(6-Bromo-3-ethoxy-2-fluorophenyl)pentan-2-ol ≥95%
2-(6-Bromo-3-ethoxy-2-fluorophenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₃H₁₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34882020
CAS Number: 2969398-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR