(2-Ethoxyphenyl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

278679-1G 278679-500MG 278679-5G
77468-028EA 576.8 USD
77468-028 77480-722 77491-074
(2-Ethoxyphenyl)phenylmethanone ≥95%
(2-Ethoxyphenyl)phenylmethanone

Precaución: For research use only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01123314
CAS Number: 127154-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR