[5-(1,1-Dimethylethyl)-2-methoxyphenyl]phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

278641-1G 278641-500MG 278641-5G 278641-25G
77468-024EA 415.09 USD
77468-024 77480-718 77491-070 77501-108
[5-(1,1-Dimethylethyl)-2-methoxyphenyl]phenylmethanone ≥95%
[5-(1,1-Dimethylethyl)-2-methoxyphenyl]phenylmethanone

Precaución: For research use only

Formula: C₁₈H₂₀O₂
MW: 268.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19303746
CAS Number: 5409-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR