3'-Bromo[1,1'-biphenyl]-4-amine ≥95%

Supplier: AOBChem USA

Warning

278494-1G 278494-500MG 278494-5G
77468-020EA 504.7 USD
77468-020 77480-714 77491-066
3'-Bromo[1,1'-biphenyl]-4-amine ≥95%
3'-Bromo[1,1'-biphenyl]-4-amine

Precaución: For research use only

Formula: C₁₂H₁₀BrN
MW: 248.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11574754
CAS Number: 91394-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR