2,6-Dibromopyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

27804-1G 27804-500MG 27804-5G
77468-016EA 519.12 USD
77468-016 77480-710 77491-062
2,6-Dibromopyridine-4-carboxaldehyde ≥97%
2,6-Dibromopyridine-4-carboxaldehyde

Precaución: For research use only

Formula: C₆H₃Br₂NO
MW: 264.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410666
CAS Number: 316800-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR