α-[2-(Trimethylsilyl)ethynyl]cyclohexanemethanol ≥95%

Supplier: AOBChem USA

Warning

277692-1G 277692-500MG 277692-5G
77468-012EA 451.14 USD
77468-012 77480-706 77491-058
α-[2-(Trimethylsilyl)ethynyl]cyclohexanemethanol ≥95%
α-[2-(Trimethylsilyl)ethynyl]cyclohexanemethanol

Precaución: For research use only

Formula: C₁₂H₂₂OSi
MW: 210.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34881804
CAS Number: 112497-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR