4-Methyl-1-(trimethylsilyl)-1-pentyn-3-ol ≥95%

Supplier: AOBChem USA

Warning

277652-1G 277652-500MG 277652-5G 277652-25G
77468-008EA 390.37 USD
77468-008 77480-702 77491-054 77501-102
4-Methyl-1-(trimethylsilyl)-1-pentyn-3-ol ≥95%
4-Methyl-1-(trimethylsilyl)-1-pentyn-3-ol

Precaución: For research use only

Formula: C₉H₁₈OSi
Storage Temperature: Ambient
MDL Number: MFCD05864337
CAS Number: 61077-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR