2-Methyl-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

277537-1G 277537-500MG 277537-5G 277537-25G
77468-004EA 390.37 USD
77468-004 77480-698 77491-050 77501-098
2-Methyl-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%
2-Methyl-α-[2-(trimethylsilyl)ethynyl]benzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈OSi
MW: 218.37 g/mol
Storage Temperature: Ambient
CAS Number: 153889-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR