3-Benzyloxy-5-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

27750-1G 27750-500MG 27750-5G 27750-25G
77468-000EA 368.74 USD
77468-000 77480-694 77491-044 77501-094
3-Benzyloxy-5-hydroxypyridine ≥97%
3-Benzyloxy-5-hydroxypyridine

Precaución: For research use only

Formula: C₁₂H₁₁NO₂
MW: 201.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20923567
CAS Number: 209328-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR