5-Methyl-2-(methylthio)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

27735-1G 27735-500MG 27735-5G
77467-996EA 519.12 USD
77467-996 77480-690 77491-040
5-Methyl-2-(methylthio)nicotinaldehyde ≥97%
5-Methyl-2-(methylthio)nicotinaldehyde

Precaución: For research use only

Formula: C₈H₉NOS
MW: 167.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142615
CAS Number: 1289211-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR