4-Phenoxy-2-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27717-1G 27717-500MG 27717-5G
77467-992EA 922.88 USD
77467-992 77480-688 77491-036
4-Phenoxy-2-(trifluoromethyl)phenylboronic acid ≥97%
4-Phenoxy-2-(trifluoromethyl)phenylboronic acid

Precaución: For research use only

Formula: C₁₃H₁₀BF₃O₃
MW: 282.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360682
CAS Number: 1415824-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR