3-Methyl-1-(4-(methylthio)phenyl)butan-1-ol ≥97%

Supplier: AOBChem USA

Warning

276433-1G 276433-500MG 276433-5G 276433-25G
77467-988EA 226.6 USD
77467-988 77480-684 77491-032 77501-084
3-Methyl-1-(4-(methylthio)phenyl)butan-1-ol ≥97%
3-Methyl-1-(4-(methylthio)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.3357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34881476
CAS Number: 1601988-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR