1-(6-Bromo-3-ethoxy-2-fluorophenyl)-2-methylpentan-1-ol ≥95%

Supplier: AOBChem USA

Warning

276430-1G 276430-500MG 276430-5G 276430-25G
77467-984EA 415.09 USD
77467-984 77480-680 77491-028 77501-080
1-(6-Bromo-3-ethoxy-2-fluorophenyl)-2-methylpentan-1-ol ≥95%
1-(6-Bromo-3-ethoxy-2-fluorophenyl)-2-methylpentan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₀BrFO₂
MW: 319.2098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34881473
CAS Number: 2969322-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR