5-Ethoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

27599-1G 27599-500MG 27599-5G 27599-25G
77467-980EA 253.38 USD
77467-980 77480-676 77491-024 77501-078
5-Ethoxy-2-methylbenzaldehyde ≥97%
5-Ethoxy-2-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142604
CAS Number: 1809157-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR