1-Ethoxy-2,3-difluoro-4-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

275434-1G 275434-500MG 275434-5G 275434-25G
77467-976EA 415.09 USD
77467-976 77480-672 77491-018 77501-070
1-Ethoxy-2,3-difluoro-4-(trimethylsilyl)benzene ≥95%
1-Ethoxy-2,3-difluoro-4-(trimethylsilyl)benzene

Precaución: For research use only

Formula: C₁₁H₁₆F₂OSi
MW: 230.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34881196
CAS Number: 138871-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR