1-(5-Bromo-3-ethoxy-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

275366-1G 275366-500MG 275366-5G 275366-25G
77467-972EA 415.09 USD
77467-972 77480-668 77491-014 77501-068
1-(5-Bromo-3-ethoxy-2-fluorophenyl)ethanone ≥95%
1-(5-Bromo-3-ethoxy-2-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34881157
CAS Number: 2385823-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR