Thymyl acetate ≥96%

Supplier: AOBChem USA

Warning

274973-1G 274973-500MG 274973-5G 274973-25G
77467-968EA 304.88 USD
77467-968 77480-664 77491-010 77501-064
Thymyl acetate ≥96%
Thymyl acetate

Precaución: For research use only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD02149365
CAS Number: 528-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR