4-Chloro-α-[3-(1-methylethoxy)phenyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

274669-1G 274669-500MG 274669-5G 274669-25G
77467-964EA 415.09 USD
77467-964 77480-660 77491-006 77501-058
4-Chloro-α-[3-(1-methylethoxy)phenyl]benzenemethanol ≥95%
4-Chloro-α-[3-(1-methylethoxy)phenyl]benzenemethanol

Precaución: For research use only

Formula: C₁₆H₁₇ClO₂
MW: 276.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17404249
CAS Number: 1271595-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR