Benzenemethanol, 4-butyl-α-(4-chlorophenyl)- ≥95%

Supplier: AOBChem USA

Warning

274658-1G 274658-500MG 274658-5G 274658-25G
77467-960EA 415.09 USD
77467-960 77480-656 77491-002 77501-054
Benzenemethanol, 4-butyl-α-(4-chlorophenyl)- ≥95%
Benzenemethanol, 4-butyl-α-(4-chlorophenyl)-

Precaución: For research use only

Formula: C₁₇H₁₉ClO
MW: 274.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11097023
CAS Number: 944698-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR