α-(3-Chlorophenyl)-2,5-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

274573-1G 274573-500MG 274573-5G 274573-25G
77467-956EA 415.09 USD
77467-956 77480-652 77490-998 77501-050
α-(3-Chlorophenyl)-2,5-dimethoxybenzenemethanol ≥95%
α-(3-Chlorophenyl)-2,5-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₅H₁₅ClO₃
MW: 278.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16357113
CAS Number: 1225863-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR