α-(3-Chlorophenyl)[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

274537-1G 274537-500MG 274537-5G 274537-25G
77467-952EA 415.09 USD
77467-952 77480-648 77490-994 77501-046
α-(3-Chlorophenyl)[1,1'-biphenyl]-4-methanol ≥95%
α-(3-Chlorophenyl)[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₉H₁₅ClO
MW: 294.78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936740
CAS Number: 1406297-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR