1-[3-Bromo-5-(trifluoromethyl)phenyl]-N,N-dimethylmethanamine ≥97%

Supplier: AOBChem USA

Warning

27425-1G 27425-500MG 27425-5G 27425-25G
77467-948EA 258.53 USD
77467-948 77480-644 77490-990 77501-042
1-[3-Bromo-5-(trifluoromethyl)phenyl]-N,N-dimethylmethanamine ≥97%
1-[3-Bromo-5-(trifluoromethyl)phenyl]-N,N-dimethylmethanamine

Precaución: For research use only

Formula: C₁₀H₁₁BrF₃N
MW: 282.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30536503
CAS Number: 1237535-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR