1-Methyl-4-(phenylethynyl)piperidin-4-ol ≥95%

Supplier: AOBChem USA

Warning

274191-1G 274191-500MG 274191-5G
77467-940EA 546.93 USD
77467-940 77480-636 77490-984
1-Methyl-4-(phenylethynyl)piperidin-4-ol ≥95%
1-Methyl-4-(phenylethynyl)piperidin-4-ol

Precaución: For research use only

Formula: C₁₄H₁₇NO
MW: 215.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34880873
CAS Number: 54848-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR