2-Nitrophenyl acetate ≥97%

Supplier: AOBChem USA

Warning

273808-1G 273808-500MG 273808-5G 273808-25G
77467-936EA 106.09 USD
77467-936 77480-632 77490-980 77501-032
2-Nitrophenyl acetate ≥97%
2-Nitrophenyl acetate

Precaución: For research use only

Formula: C₈H₇NO₄
MW: 181.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007095
CAS Number: 610-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR