3-Methyl-1-(3-(methylthio)phenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

273778-1G 273778-500MG 273778-5G
77467-932EA 434.66 USD
77467-932 77480-628 77490-976
3-Methyl-1-(3-(methylthio)phenyl)butan-1-ol ≥95%
3-Methyl-1-(3-(methylthio)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.3357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34880665
CAS Number: 2152237-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR